siRNA Details
Gene Symbol: Zfp593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106689.1 | NP_001100159.1 | 3 | 447 | ||
GenBank mRNA | BC166512.1 | - | 3 | 403 | 7128768 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106689.1 | NP_001100159.1 | 3 | 447 | ||
GenBank mRNA | BC166512.1 | - | 3 | 403 | 7128768 |