siRNA Details
| Gene Symbol: Pithd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_216543.7 | XP_216543.5 | 4 | 497 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pithd1| Gene Symbol: Pithd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_216543.7 | XP_216543.5 | 4 | 497 | ||