siRNA Details
| Gene Symbol: Pramef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238356.2 | XP_006238418.1 | 3 | 1017 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pramef5| Gene Symbol: Pramef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238356.2 | XP_006238418.1 | 3 | 1017 | ||