siRNA Details
| Gene Symbol: Ccnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013094.1 | NP_001013112.1 | 5, 6 | 645 | ||
| GenBank mRNA | BC088316.1 | - | 5, 6 | 645 | 7325177 | |
| FQ211043.1 | - | 5, 6 | 706 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccnl2| Gene Symbol: Ccnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013094.1 | NP_001013112.1 | 5, 6 | 645 | ||
| GenBank mRNA | BC088316.1 | - | 5, 6 | 645 | 7325177 | |
| FQ211043.1 | - | 5, 6 | 706 | |||