siRNA Details
| Gene Symbol: Ube2j2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007655.1 | NP_001007656.1 | 4 | 642 | ||
| XM_006239554.2 | XP_006239616.1 | 4 | 494 | |||
| XM_006239555.2 | XP_006239617.1 | 5 | 829 | |||
| XM_006239557.2 | XP_006239619.1 | 3 | 489 | |||
| XM_006239558.2 | XP_006239620.1 | 3 | 331 | |||
| XM_008764352.1 | XP_008762574.1 | 4 | 652 | |||
| XM_008764353.1 | XP_008762575.1 | 4 | 1722 | |||
| XM_008764354.1 | XP_008762576.1 | 5 | 833 | |||
| XM_008764355.1 | XP_008762577.1 | 5 | 1778 | |||
| GenBank mRNA | BC079115.1 | - | 4 | 642 | 7126201 | |

Ube2j2
Genomic Map