siRNA Details
Gene Symbol: Isg15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106700.1 | NP_001100170.1 | 2 | 295 | ||
XM_006239563.2 | XP_006239625.1 | 3 | 889 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Isg15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106700.1 | NP_001100170.1 | 2 | 295 | ||
XM_006239563.2 | XP_006239625.1 | 3 | 889 |