siRNA Details
| Gene Symbol: Isg15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106700.1 | NP_001100170.1 | 2 | 585 | ||
| XM_006239563.2 | XP_006239625.1 | 3 | 1179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Isg15| Gene Symbol: Isg15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106700.1 | NP_001100170.1 | 2 | 585 | ||
| XM_006239563.2 | XP_006239625.1 | 3 | 1179 | |||