siRNA Details
| Gene Symbol: Raver1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013939.1 | NP_001013961.1 | 3 | 496 | ||
| GenBank mRNA | BC083865.1 | - | 3 | 496 | 7127766 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Raver1| Gene Symbol: Raver1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013939.1 | NP_001013961.1 | 3 | 496 | ||
| GenBank mRNA | BC083865.1 | - | 3 | 496 | 7127766 | |