siRNA Details
| Gene Symbol: Ypel5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035221.1 | NP_001030298.1 | 461 | |||
| XM_008764508.1 | XP_008762730.1 | 668 | ||||
| XM_008764509.1 | XP_008762731.1 | 916 | ||||
| XM_008764510.1 | XP_008762732.1 | 867 | ||||
| GenBank mRNA | CB584328.1 | - | 472 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_027166, AK046281, AK049644, AK169986, AK167998, and others.

Ypel5
Genomic Map