siRNA Details
| Gene Symbol: Clip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013942.1 | NP_001013964.1 | 12, 13 | 1648 | ||
| GenBank mRNA | BC081910.1 | - | 12, 13 | 1648 | 7111548 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clip4| Gene Symbol: Clip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013942.1 | NP_001013964.1 | 12, 13 | 1648 | ||
| GenBank mRNA | BC081910.1 | - | 12, 13 | 1648 | 7111548 | |