siRNA Details
| Gene Symbol: Cenpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106711.1 | NP_001100181.1 | 2, 3 | 290 | ||
| XM_006239798.2 | XP_006239860.1 | 2, 3 | 413 | |||
| GenBank mRNA | BC166997.1 | - | 2, 3 | 331 | 8365241 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cenpa| Gene Symbol: Cenpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106711.1 | NP_001100181.1 | 2, 3 | 290 | ||
| XM_006239798.2 | XP_006239860.1 | 2, 3 | 413 | |||
| GenBank mRNA | BC166997.1 | - | 2, 3 | 331 | 8365241 | |