siRNA Details
| Gene Symbol: Gpr113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106712.1 | NP_001100182.1 | 9 | 2577 | ||
| XM_006239800.2 | XP_006239862.1 | 10 | 2850 | |||
| XM_006239801.2 | XP_006239863.1 | 9 | 2709 | |||
More Information
Other Species Targeted
Set Membership: |
|

Gpr113
Genomic Map