siRNA Details
| Gene Symbol: Slc7a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106714.1 | NP_001100184.1 | 1 | 485 | ||
| XM_008764578.1 | XP_008762800.1 | 1 | 775 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc7a15| Gene Symbol: Slc7a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106714.1 | NP_001100184.1 | 1 | 485 | ||
| XM_008764578.1 | XP_008762800.1 | 1 | 775 | |||