siRNA Details
| Gene Symbol: Laptm4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199384.2 | NP_955416.1 | 2 | 275 | ||
| GenBank mRNA | BC063179.1 | - | 2 | 279 | 6890564 | |
| FQ212732.1 | - | 2 | 677 | |||
| FQ213363.1 | - | 2 | 309 | |||
| FQ229677.1 | - | 234 | ||||
| FQ232171.1 | - | 2 | 333 | |||
| FQ232930.1 | - | 2 | 293 | |||
| FQ235113.1 | - | 2 | 180 | |||

Laptm4a
Genomic Map