siRNA Details
| Gene Symbol: Laptm4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199384.2 | NP_955416.1 | 2, 3 | 310 | ||
| GenBank mRNA | BC063179.1 | - | 2, 3 | 314 | 6890564 | |
| FQ212732.1 | - | 2, 3 | 712 | |||
| FQ213363.1 | - | 2, 3 | 344 | |||
| FQ229677.1 | - | 269 | ||||
| FQ232171.1 | - | 2, 3 | 368 | |||
| FQ232930.1 | - | 2, 3 | 328 | |||
| FQ235113.1 | - | 2, 3 | 215 | |||

Laptm4a
Genomic Map