siRNA Details
| Gene Symbol: Pqlc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034952.1 | NP_001030124.1 | 5, 6 | 640 | ||
| GenBank mRNA | CK477351.1 | - | 5, 6 | 463 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pqlc3| Gene Symbol: Pqlc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034952.1 | NP_001030124.1 | 5, 6 | 640 | ||
| GenBank mRNA | CK477351.1 | - | 5, 6 | 463 | ||