siRNA Details
Gene Symbol: Dld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199385.2 | NP_955417.1 | 10 | 1059 | ||
XM_008764590.1 | XP_008762812.1 | 10 | 1076 | |||
GenBank mRNA | BC062069.1 | - | 10 | 1063 | 5599215 | |
FM046215.1 | - | 2 | 229 | |||
FQ215299.1 | - | 10 | 1066 |
