siRNA Details
| Gene Symbol: Dld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199385.2 | NP_955417.1 | 7 | 538 | ||
| XM_008764590.1 | XP_008762812.1 | 7 | 555 | |||
| GenBank mRNA | BC062069.1 | - | 7 | 542 | 5599215 | |
| FM093110.1 | - | 2 | 116 | |||
| FM100976.1 | - | 7 | 538 | |||
| FQ215299.1 | - | 7 | 545 | |||

Dld
Genomic Map