siRNA Details
Gene Symbol: Bcap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006980.1 | NP_001006981.1 | 3 | 235 | ||
XM_006239982.2 | XP_006240044.1 | 4 | 356 | |||
GenBank mRNA | BC083576.1 | - | 3 | 235 | 7113186 |
