siRNA Details
Gene Symbol: Bcap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006980.1 | NP_001006981.1 | 6 | 660 | ||
XM_006239982.2 | XP_006240044.1 | 7 | 781 | |||
GenBank mRNA | BC083576.1 | - | 6 | 660 | 7113186 |
