siRNA Details
| Gene Symbol: Sav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001097581.1 | NP_001091050.1 | 5 | 1193 | ||
| GenBank mRNA | AM261215.1 | - | 5 | 1193 | ||
| BC161803.1 | - | 5 | 1040 | 8371277 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sav1| Gene Symbol: Sav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001097581.1 | NP_001091050.1 | 5 | 1193 | ||
| GenBank mRNA | AM261215.1 | - | 5 | 1193 | ||
| BC161803.1 | - | 5 | 1040 | 8371277 | ||