siRNA Details
Gene Symbol: Dhrs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013098.1 | NP_001013116.1 | 4 | 532 | ||
GenBank mRNA | AB108671.1 | - | 4 | 532 | ||
BC104713.1 | - | 4 | 438 | 7370181 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dhrs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013098.1 | NP_001013116.1 | 4 | 532 | ||
GenBank mRNA | AB108671.1 | - | 4 | 532 | ||
BC104713.1 | - | 4 | 438 | 7370181 |