siRNA Details
| Gene Symbol: Rhoj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008320.1 | NP_001008321.1 | 3, 4 | 658 | ||
| GenBank mRNA | BC086525.1 | - | 3, 4 | 658 | 7303905 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhoj| Gene Symbol: Rhoj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008320.1 | NP_001008321.1 | 3, 4 | 658 | ||
| GenBank mRNA | BC086525.1 | - | 3, 4 | 658 | 7303905 | |