siRNA Details
Gene Symbol: Rab15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198749.2 | NP_942044.1 | 1 | 399 | ||
GenBank mRNA | BC094954.1 | - | 1 | 399 | 7316003 | |
M83679.1 | - | 1 | 243 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rab15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198749.2 | NP_942044.1 | 1 | 399 | ||
GenBank mRNA | BC094954.1 | - | 1 | 399 | 7316003 | |
M83679.1 | - | 1 | 243 |