siRNA Details
| Gene Symbol: Ift43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134525.1 | NP_001127997.1 | 2 | 143 | ||
| XM_006240370.2 | XP_006240432.1 | 2 | 129 | |||
| XM_006240372.2 | XP_006240434.1 | 2 | 129 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ift43| Gene Symbol: Ift43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134525.1 | NP_001127997.1 | 2 | 143 | ||
| XM_006240370.2 | XP_006240432.1 | 2 | 129 | |||
| XM_006240372.2 | XP_006240434.1 | 2 | 129 | |||