siRNA Details
Gene Symbol: Zdhhc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039325.1 | NP_001034414.1 | 2 | 512 | ||
GenBank mRNA | AY886537.1 | - | 2 | 512 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zdhhc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039325.1 | NP_001034414.1 | 2 | 512 | ||
GenBank mRNA | AY886537.1 | - | 2 | 512 |