siRNA Details
| Gene Symbol: Zdhhc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039325.1 | NP_001034414.1 | 3 | 822 | ||
| GenBank mRNA | AY886537.1 | - | 3 | 822 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zdhhc22| Gene Symbol: Zdhhc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039325.1 | NP_001034414.1 | 3 | 822 | ||
| GenBank mRNA | AY886537.1 | - | 3 | 822 | ||