siRNA Details
| Gene Symbol: Ifi27l2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206846.2 | NP_996728.1 | 3 | 170 | ||
| GenBank mRNA | BC166870.1 | - | 3 | 170 | 6922110 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifi27l2b| Gene Symbol: Ifi27l2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206846.2 | NP_996728.1 | 3 | 170 | ||
| GenBank mRNA | BC166870.1 | - | 3 | 170 | 6922110 | |