siRNA Details
| Gene Symbol: Ndufb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106756.1 | NP_001100226.1 | 1 | 437 | ||
| GenBank mRNA | FQ211881.1 | - | 1 | 152 | ||
| FQ217020.1 | - | 1 | 153 | |||
| FQ217041.1 | - | 1 | 151 | |||
| FQ217852.1 | - | 1 | 157 | |||
| FQ223751.1 | - | 1 | 155 | |||
| FQ224251.1 | - | 1 | 152 | |||
| FQ224299.1 | - | 1 | 153 | |||
| FQ224538.1 | - | 1 | 166 | |||

Ndufb11
Genomic Map