siRNA Details
| Gene Symbol: Ndufb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106756.1 | NP_001100226.1 | 3 | 657 | ||
| GenBank mRNA | FQ211881.1 | - | 3 | 372 | ||
| FQ217020.1 | - | 3 | 373 | |||
| FQ217041.1 | - | 3 | 371 | |||
| FQ217852.1 | - | 3 | 377 | |||
| FQ223751.1 | - | 3 | 375 | |||
| FQ223793.1 | - | 3 | 403 | |||
| FQ224251.1 | - | 3 | 372 | |||
| FQ224299.1 | - | 3 | 373 | |||
| FQ224538.1 | - | 3 | 386 | |||

Ndufb11
Genomic Map