siRNA Details
Gene Symbol: Hsp90aa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175761.2 | NP_786937.1 | 3 | 392 | ||
GenBank mRNA | AJ428213.1 | - | 3 | 392 | ||
AY683455.1 | - | 2 | 233 | |||
BC072489.1 | - | 3 | 374 | 7105514 | ||
BC085120.1 | - | 3 | 374 | 7114546 | ||
FQ211921.1 | - | 3 | 392 | |||
FQ213634.1 | - | 3 | 391 | |||
FQ225235.1 | - | 3 | 548 | |||
FQ225422.1 | - | 3 | 392 | |||
FQ225565.1 | - | 3 | 392 | |||
FQ225903.1 | - | 3 | 548 | |||
FQ225992.1 | - | 3 | 392 | |||
FQ226310.1 | - | 3 | 392 | |||
FQ226483.1 | - | 3 | 392 | |||
FQ226612.1 | - | 3 | 393 | |||
FQ226980.1 | - | 3 | 392 | |||
FQ227103.1 | - | 3 | 392 | |||
FQ227479.1 | - | 3 | 392 | |||
FQ227490.1 | - | 3 | 391 | |||
FQ227525.1 | - | 3 | 393 | |||
FQ228040.1 | - | 3 | 392 | |||
FQ230194.1 | - | 3 | 393 | |||
FQ230221.1 | - | 3 | 392 | |||
FQ230359.1 | - | 3 | 392 | |||
FQ230408.1 | - | 3 | 392 | |||
FQ230565.1 | - | 3 | 392 | |||
FQ230623.1 | - | 3 | 392 | |||
FQ231096.1 | - | 3 | 392 | |||
FQ231403.1 | - | 3 | 392 | |||
FQ231559.1 | - | 3 | 396 | |||
FQ231773.1 | - | 3 | 468 | |||
FQ232301.1 | - | 3 | 548 | |||
FQ232340.1 | - | 3 | 392 | |||
FQ232738.1 | - | 3 | 392 | |||
FQ232984.1 | - | 3 | 392 | |||
FQ233436.1 | - | 3 | 392 | |||
FQ233585.1 | - | 3 | 392 | |||
FQ234090.1 | - | 3 | 392 | |||
FQ234628.1 | - | 3 | 392 | |||
FQ235318.1 | - | 3 | 392 |
Gene Symbol: LOC103692716 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764969.1 | XP_008763191.1 | 444 |
