siRNA Details
| Gene Symbol: Cinp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106758.2 | NP_001100228.1 | 3, 4 | 330 | ||
| GenBank mRNA | BC158883.1 | - | 3, 4 | 330 | 7937483 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cinp| Gene Symbol: Cinp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106758.2 | NP_001100228.1 | 3, 4 | 330 | ||
| GenBank mRNA | BC158883.1 | - | 3, 4 | 330 | 7937483 | |