siRNA Details
| Gene Symbol: Cyp4f39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241090.1 | XP_006241152.1 | 8 | 964 | ||
| XM_006241091.2 | XP_006241153.1 | 8 | 1073 | |||
| XM_006241092.1 | XP_006241154.1 | 7 | 876 | |||
| XM_008765183.1 | XP_008763405.1 | 7 | 1304 | |||
| XM_234837.7 | XP_234837.3 | 8 | 1405 | |||
| GenBank mRNA | FQ210490.1 | - | 8 | 969 | ||

Cyp4f39
Genomic Map