siRNA Details
| Gene Symbol: Olr1088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765192.1 | XP_008763414.1 | 1 | 284 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1088| Gene Symbol: Olr1088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765192.1 | XP_008763414.1 | 1 | 284 | ||