siRNA Details
| Gene Symbol: RGD1359127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007657.1 | NP_001007658.1 | 2 | 379 | ||
| GenBank mRNA | BC079167.1 | - | 2 | 379 | 7129744 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1359127| Gene Symbol: RGD1359127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007657.1 | NP_001007658.1 | 2 | 379 | ||
| GenBank mRNA | BC079167.1 | - | 2 | 379 | 7129744 | |