siRNA Details
| Gene Symbol: Lmnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765148.1 | XP_008763370.1 | 3 | 1129 | ||
| XM_008765149.1 | XP_008763371.1 | 3 | 1400 | |||
| XM_216850.8 | XP_216850.6 | 2 | 1541 | |||
| GenBank mRNA | FQ212316.1 | - | 2 | 365 | ||

Lmnb2
Genomic Map