siRNA Details
| Gene Symbol: Mrpl54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106770.1 | NP_001100240.1 | 2 | 301 | ||
| GenBank mRNA | FQ211295.1 | - | 2 | 294 | ||
| FQ211839.1 | - | 2 | 284 | |||
| FQ217217.1 | - | 2 | 284 | |||
| FQ221492.1 | - | 2 | 288 | |||
| FQ224331.1 | - | 2 | 288 | |||
| FQ224393.1 | - | 2 | 284 | |||
| FQ224740.1 | - | 2 | 286 | |||

Mrpl54
Genomic Map