siRNA Details
Gene Symbol: Ano4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106778.1 | NP_001100248.1 | 11 | 1545 | ||
XM_008765227.1 | XP_008763449.1 | 8 | 6704 | |||
XM_008765228.1 | XP_008763450.1 | 8 | 739 | |||
XM_008765229.1 | XP_008763451.1 | 7 | 6659 | |||
XM_008765230.1 | XP_008763452.1 | 7 | 634 | |||
XM_008765231.1 | XP_008763453.1 | 12 | 2372 |
