siRNA Details
| Gene Symbol: Ndufa12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106781.2 | NP_001100251.2 | 4 | 308 | ||
| GenBank mRNA | BF555697.1 | - | 4 | 283 | ||
| BG671355.1 | - | 3 | 159 | |||
| BQ205274.1 | - | 256 | ||||
| FQ212338.1 | - | 4 | 293 | |||
| FQ217295.1 | - | 4 | 294 | |||
| FQ224477.1 | - | 4 | 294 | |||

Ndufa12
Genomic Map