siRNA Details
| Gene Symbol: Ndufa12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106781.2 | NP_001100251.2 | 4 | 434 | ||
| GenBank mRNA | BF555697.1 | - | 4 | 409 | ||
| BG671355.1 | - | 3 | 285 | |||
| BQ205274.1 | - | 130 | ||||
| FQ212338.1 | - | 4 | 419 | |||
| FQ217295.1 | - | 4 | 420 | |||
| FQ224477.1 | - | 4 | 420 | |||

Ndufa12
Genomic Map