siRNA Details
| Gene Symbol: Cpsf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106785.1 | NP_001100255.1 | 4 | 504 | ||
| XM_006241372.1 | XP_006241434.1 | 4 | 536 | |||
| XM_006241373.1 | XP_006241435.1 | 4 | 536 | |||
| XM_006241374.1 | XP_006241436.1 | 4 | 536 | |||
| XM_008765386.1 | XP_008763608.1 | 4 | 536 | |||
| XM_008765387.1 | XP_008763609.1 | 4 | 536 | |||
| XM_008765388.1 | XP_008763610.1 | 4 | 536 | |||
| XM_008765390.1 | XP_008763612.1 | 4 | 461 | |||

Cpsf6
Genomic Map