siRNA Details
| Gene Symbol: Tmem5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024759.1 | NP_001019930.1 | 4 | 771 | ||
| XM_006241397.2 | XP_006241459.1 | 4 | 836 | |||
| GenBank mRNA | BC097355.1 | - | 4 | 771 | 7441970 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem5| Gene Symbol: Tmem5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024759.1 | NP_001019930.1 | 4 | 771 | ||
| XM_006241397.2 | XP_006241459.1 | 4 | 836 | |||
| GenBank mRNA | BC097355.1 | - | 4 | 771 | 7441970 | |