siRNA Details
| Gene Symbol: LOC103690326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765471.1 | XP_008763693.1 | 1586 | |||
| XM_008765472.1 | XP_008763694.1 | 1586 | ||||
| XM_008765473.1 | XP_008763695.1 | 1586 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103690326| Gene Symbol: LOC103690326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765471.1 | XP_008763693.1 | 1586 | |||
| XM_008765472.1 | XP_008763694.1 | 1586 | ||||
| XM_008765473.1 | XP_008763695.1 | 1586 | ||||