siRNA Details
Gene Symbol: Eif3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198751.1 | NP_942046.1 | 2 | 430 | ||
GenBank mRNA | BC060586.1 | - | 2 | 430 | 6918037 | |
FQ216838.1 | - | 2 | 427 | |||
FQ220633.1 | - | 2 | 429 | |||
FQ228544.1 | - | 2 | 413 | |||
FQ229203.1 | - | 2 | 423 | |||
FQ232428.1 | - | 2 | 428 |
Gene Symbol: LOC100911110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750101.3 | XP_003750149.1 | 3 | 430 |
