siRNA Details
Gene Symbol: Ndrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011991.1 | NP_001011991.1 | 6 | 491 | ||
XM_006241708.2 | XP_006241770.1 | 6 | 763 | |||
GenBank mRNA | AY500369.1 | - | 6 | 453 | ||
BC081898.1 | - | 6 | 491 | 7110455 | ||
FQ212154.1 | - | 6 | 503 |
