siRNA Details
| Gene Symbol: Lypd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130545.1 | NP_001124017.1 | 2, 3 | 250 | ||
| XM_006241726.2 | XP_006241788.1 | 2, 3 | 438 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lypd2| Gene Symbol: Lypd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130545.1 | NP_001124017.1 | 2, 3 | 250 | ||
| XM_006241726.2 | XP_006241788.1 | 2, 3 | 438 | |||