siRNA Details
| Gene Symbol: Eif3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034134.1 | NP_001029306.1 | 3 | 510 | ||
| GenBank mRNA | BC100264.1 | - | 3 | 510 | 7382342 | |
| FQ209474.1 | - | 535 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif3l| Gene Symbol: Eif3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034134.1 | NP_001029306.1 | 3 | 510 | ||
| GenBank mRNA | BC100264.1 | - | 3 | 510 | 7382342 | |
| FQ209474.1 | - | 535 | ||||