siRNA Details
| Gene Symbol: Phf21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130680.1 | NP_001124152.1 | 2, 3 | 521 | ||
| XM_008765685.1 | XP_008763907.1 | 2, 3 | 108 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phf21b| Gene Symbol: Phf21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130680.1 | NP_001124152.1 | 2, 3 | 521 | ||
| XM_008765685.1 | XP_008763907.1 | 2, 3 | 108 | |||