siRNA Details
| Gene Symbol: Smc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130498.1 | NP_001123970.1 | 19 | 2917 | ||
| XM_008765687.1 | XP_008763909.1 | 19 | 2952 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smc1b| Gene Symbol: Smc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130498.1 | NP_001123970.1 | 19 | 2917 | ||
| XM_008765687.1 | XP_008763909.1 | 19 | 2952 | |||