siRNA Details
Gene Symbol: Ttc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130499.1 | NP_001123971.1 | 13 | 1328 | ||
GenBank mRNA | FQ210197.1 | - | 13 | 1297 | ||
FQ218771.1 | - | 13 | 1338 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ttc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130499.1 | NP_001123971.1 | 13 | 1328 | ||
GenBank mRNA | FQ210197.1 | - | 13 | 1297 | ||
FQ218771.1 | - | 13 | 1338 |